5,5-Dimethyl-2-(3-methylphenyl)-1,3,2-dioxaborinane 95+%

Supplier: Ambeed
Warning

A441029-250MG A441029-5G
77156-056EA 8 USD
77156-056 77156-058
5,5-Dimethyl-2-(3-methylphenyl)-1,3,2-dioxaborinane 95+%
5,5-Dimethyl-2-(3-methylphenyl)-1,3,2-dioxaborinane

Precaución: for Research Use Only

Formula: C₁₂H₁₇BO₂
MDL Number: MFCD28101508
CAS Number: 223799-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR