RITA 98%

Supplier: Ambeed
Warning

A440096-10MG A440096-1MG A440096-2MG A440096-5MG A440096-1G A440096-250MG A440096-100MG
77156-042EA 74 USD
77156-042 77156-044 77156-046 77156-048 77686-192 77686-193 77901-581
RITA 98%
RITA

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₃S₂
MDL Number: MFCD03235294
CAS Number: 213261-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR