2,2,2,2'-Tetrafluoroacetophenone 97%

Supplier: Ambeed
Danger

A439922-250MG A439922-1G
77156-040EA 20 USD
77156-040 77900-797
2,2,2,2'-Tetrafluoroacetophenone 97%
2,2,2,2'-Tetrafluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₄F₄O
MDL Number: MFCD00040099
CAS Number: 124004-75-7
UN: 1224
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR