8-Bromo-4-hydroxy-2-methylquinoline 98%

Supplier: Ambeed
Warning

Synonyms: 8-Bromo-2-methylquinolin-4-ol

A439527-100MG A439527-1G A439527-250MG A439527-5G
77156-032EA 20.6 USD
77156-032 77156-034 77156-036 77156-038
8-Bromo-4-hydroxy-2-methylquinoline 98%
8-Bromo-4-hydroxy-2-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO
MW: 238.081 g/mol
MDL Number: MFCD00484434
CAS Number: 1201-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR