1-Hexyl-3-methylimidazolium bromide 98%

Supplier: Ambeed
Warning

A438865-100G A438865-1G A438865-25G A438865-5G
77156-016EA 94.76 USD
77156-016 77156-018 77156-020 77156-022
1-Hexyl-3-methylimidazolium bromide 98%
1-Hexyl-3-methylimidazolium bromide

Precaución: for Research Use Only

Formula: C₁₀H₁₉BrN₂
MW: 247.18 g/mol
MDL Number: MFCD03427612
CAS Number: 85100-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR