Methyl 3-fluoro-5-formylbenzoate 97%
Supplier: Ambeed
|
Warning
|
A438590-100MG
A438590-1G
A438590-250MG
A438590-50MG
A438590-5G
77155-998EA
149.35
USD
77155-998
77156-000
77156-002
77156-004
77156-006
Methyl 3-fluoro-5-formylbenzoate 97%
Methyl 3-fluoro-5-formylbenzoate
Precaución: for Research Use Only
Formula:
C₉H₇FO₃ MW: 182.148 g/mol |
MDL Number:
MFCD22544332 CAS Number: 1393561-99-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...