1-Bromo-4,5-dichloro-2-fluorobenzene 97%

Supplier: Ambeed
Warning

A438440-100MG A438440-1G A438440-5G A438440-250MG
77155-992EA 16 USD
77155-992 77155-994 77155-996 77901-439
1-Bromo-4,5-dichloro-2-fluorobenzene 97%
1-Bromo-4,5-dichloro-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂F
MDL Number: MFCD09878158
CAS Number: 1000572-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR