4-Iodo-2,7-naphthyridin-1(2H)-one 95%

Supplier: Ambeed
Warning

A438245-100MG A438245-250MG A438245-5G
77155-988EA 24 USD
77155-988 77155-990 77892-907
4-Iodo-2,7-naphthyridin-1(2H)-one 95%
4-Iodo-2,7-naphthyridin-1(2H)-one

Precaución: for Research Use Only

Formula: C₈H₅IN₂O
MDL Number: MFCD17015944
CAS Number: 959558-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR