(R)-3-((tert-Butoxycarbonyl)amino)-4-(2,4-difluorophenyl)butanoic acid 95%

Supplier: Ambeed
Warning

A436169-1G A436169-250MG A436169-5G
77155-972EA 614.91 USD
77155-972 77155-974 77155-976
(R)-3-((tert-Butoxycarbonyl)amino)-4-(2,4-difluorophenyl)butanoic acid 95%
(R)-3-((tert-Butoxycarbonyl)amino)-4-(2,4-difluorophenyl)butanoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₉F₂NO₄
MW: 315.31 g/mol
MDL Number: MFCD27951984
CAS Number: 851307-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR