5-Chloro-1H-indole-6-carboxylic acid 98%

Supplier: Ambeed
Warning

A434963-100MG A434963-1G A434963-250MG
77155-962EA 84.46 USD
77155-962 77155-964 77155-966
5-Chloro-1H-indole-6-carboxylic acid 98%
5-Chloro-1H-indole-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆ClNO₂
MW: 195.60 g/mol
MDL Number: MFCD16875715
CAS Number: 1260858-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR