N-(2,2-Dimethylpropanoyl)-4-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-(Trifluoromethyl)phenyl)pivalamide, 4'-(Trifluoromethyl)pivalanilide

A434785-10G A434785-1G A434785-25G A434785-5G
77155-954EA 15.45 USD
77155-954 77155-956 77155-958 77155-960
N-(2,2-Dimethylpropanoyl)-4-(trifluoromethyl)aniline 98%
N-(2,2-Dimethylpropanoyl)-4-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₁₂H₁₄F₃NO
MW: 245.241 g/mol
MDL Number: MFCD03094126
CAS Number: 25617-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR