Benzo[b]thiophen-5-ylboronic acid 98%

Supplier: Ambeed
Warning

A434283-1G A434283-250MG A434283-5G
77155-948EA 154 USD
77155-948 77155-950 77155-952
Benzo[b]thiophen-5-ylboronic acid 98%
Benzo[b]thiophen-5-ylboronic acid

Precaución: for Research Use Only

Formula: C₈H₇BO₂S
MDL Number: MFCD06740319
CAS Number: 845872-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR