N-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide 97%
Supplier: Ambeed
|
Warning
|
A433236-100MG
A433236-1G
A433236-250MG
77155-942EA
30.9
USD
77155-942
77155-944
77155-946
N-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide 97%
N-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide
Precaución: for Research Use Only
Formula:
C₁₃H₁₉BN₂O₃ MW: 262.11 g/mol |
MDL Number:
MFCD11878164 CAS Number: 1313738-91-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...