7-Methoxy-4-methylquinoline hydrochloride 98%

Supplier: Ambeed
Warning

A429970-1G A429970-250MG A429970-5G
77155-910EA 50.47 USD
77155-910 77155-912 77155-914
7-Methoxy-4-methylquinoline hydrochloride 98%
7-Methoxy-4-methylquinoline hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClNO
MW: 209.67 g/mol
MDL Number: MFCD21647759
CAS Number: 1807542-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR