2,3-Dichloro-5-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Danger

A427525-10G A427525-25G A427525-5G A427525-100G A427525-500G
77155-898EA 6 USD
77155-898 77155-900 77155-902 77900-818 77900-907
2,3-Dichloro-5-(trifluoromethyl)pyridine 98%
2,3-Dichloro-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂Cl₂F₃N
MDL Number: MFCD00042243
CAS Number: 69045-84-7
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR