7-Chloro-1,2,3,4-tetrahydroquinolin-4-amine 95%

Supplier: Ambeed
Warning

A427063-100MG A427063-250MG
77155-894EA 217.33 USD
77155-894 77155-896
7-Chloro-1,2,3,4-tetrahydroquinolin-4-amine 95%
7-Chloro-1,2,3,4-tetrahydroquinolin-4-amine

Precaución: for Research Use Only

Formula: C₉H₁₁ClN₂
MW: 182.65 g/mol
MDL Number: MFCD19382231
CAS Number: 1315365-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR