1-(4-(5'-Chloro-3,5-dimethyl-[2,4'-bipyridin]-2'-yl)piperazin-1-yl)-3-(methylsulfonyl)propan-1-one 98%

Supplier: Ambeed
Warning

Synonyms: PF-5274857

A427005-10MG A427005-1MG A427005-25MG A427005-50MG A427005-5MG
77155-884EA 76.22 USD
77155-884 77155-886 77155-888 77155-890 77155-892
1-(4-(5'-Chloro-3,5-dimethyl-[2,4'-bipyridin]-2'-yl)piperazin-1-yl)-3-(methylsulfonyl)propan-1-one 98%
1-(4-(5'-Chloro-3,5-dimethyl-[2,4'-bipyridin]-2'-yl)piperazin-1-yl)-3-(methylsulfonyl)propan-1-one

Precaución: for Research Use Only

Formula: C₂₀H₂₅ClN₄O₃S
MW: 436.96 g/mol
MDL Number: MFCD22420827
CAS Number: 1373615-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR