(3R)-1-Methylpiperidin-3-amine dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (R)-3-Amino-1-methylpiperidine Dihydrochloride

A426976-10G A426976-1G A426976-250MG A426976-25G A426976-5G
77155-874EA 175 USD
77155-874 77155-876 77155-878 77155-880 77155-882
(3R)-1-Methylpiperidin-3-amine dihydrochloride 98%
(3R)-1-Methylpiperidin-3-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄N₂·2ClH
MDL Number: MFCD20926141
CAS Number: 1157849-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR