2-(tert-Butyl)isothiazol-3(2H)-one 1,1-dioxide 97+%

Supplier: Ambeed
Warning

A424542-100MG A424542-250MG
77155-856EA 50 USD
77155-856 77901-585
2-(tert-Butyl)isothiazol-3(2H)-one 1,1-dioxide 97+%
2-(tert-Butyl)isothiazol-3(2H)-one 1,1-dioxide

Precaución: for Research Use Only

Formula: C₇H₁₁NO₃S
MDL Number: MFCD28363435
CAS Number: 119639-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR