3-Dodecylthiophene 98%
Supplier: Ambeed
|
Warning
|
A424109-100G
A424109-10G
A424109-1G
A424109-25G
A424109-5G
77155-842EA
297.67
USD
77155-842
77155-844
77155-846
77155-848
77155-850
3-Dodecylthiophene 98%
3-Dodecylthiophene
Precaución: for Research Use Only
Formula:
C₁₆H₂₈S MW: 252.46 g/mol |
MDL Number:
MFCD00130121 CAS Number: 104934-52-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...