(2S,4R)-1-Boc-2-methyl-4-hydroxypiperidine 98%

Supplier: Ambeed
Warning

Synonyms: (2S,4R)-tert-Butyl 4-hydroxy-2-methylpiperidine-1-carboxylate

A423974-100MG A423974-250MG A423974-5G A423974-1G
77155-836EA 96.82 USD
77155-836 77155-838 77155-840 77594-306
(2S,4R)-1-Boc-2-methyl-4-hydroxypiperidine 98%
(2S,4R)-1-Boc-2-methyl-4-hydroxypiperidine

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₃
MW: 215.29 g/mol
MDL Number: MFCD21607787
CAS Number: 790667-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR