3-(tert-Butoxycarbonylamino)-2-methoxyisonicotinic acid 95%

Supplier: Ambeed
Warning

A423091-100MG A423091-1G A423091-250MG A423091-5G
77155-818EA 5 USD
77155-818 77155-820 77155-822 77155-824
3-(tert-Butoxycarbonylamino)-2-methoxyisonicotinic acid 95%
3-(tert-Butoxycarbonylamino)-2-methoxyisonicotinic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O₅
MDL Number: MFCD22199249
CAS Number: 870997-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR