5-Oxononanoic acid 95%

Supplier: Ambeed
Warning

A422503-1G A422503-250MG A422503-100MG
77155-812EA 309 USD
77155-812 77155-814 77901-511
5-Oxononanoic acid 95%
5-Oxononanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₆O₃
MDL Number: MFCD01320162
CAS Number: 3637-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR