Reversine 98%

Supplier: Ambeed
Warning

A418002-10MG A418002-1MG A418002-50MG A418002-5MG A418002-250MG A418002-100MG A418002-1G
77155-784EA 18 USD
77155-784 77155-786 77155-788 77155-790 77613-282 77686-156 77900-871
Reversine 98%
Reversine

Precaución: for Research Use Only

Formula: C₂₁H₂₇N₇O
MDL Number: MFCD09953189
CAS Number: 656820-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR