1-Boc-4-(3-hydroxypropyl)piperazine 98%
Supplier: Ambeed
|
Danger
|
A415813-1G
A415813-250MG
A415813-25G
A415813-5G
77155-766EA
20.6
USD
77155-766
77155-768
77155-770
77155-772
1-Boc-4-(3-hydroxypropyl)piperazine 98%
1-Boc-4-(3-hydroxypropyl)piperazine
Precaución: for Research Use Only
Formula:
C₁₂H₂₄N₂O₃ MW: 244.335 g/mol |
MDL Number:
MFCD06798090 CAS Number: 132710-90-8 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...