4-(Difluoromethyl)benzoic acid 95%

Supplier: Ambeed
Warning

A413340-250MG A413340-1G
77155-754EA 45 USD
77155-754 77572-286
4-(Difluoromethyl)benzoic acid 95%
4-(Difluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
MDL Number: MFCD07784270
CAS Number: 55805-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR