2-Fluoro-4-nitrobenzenesulfonyl chloride 97%

Supplier: Ambeed
Danger

Synonyms: 2-Fluoro-4-nitrobenzene-1-sulphonyl chloride

A413283-100MG A413283-1G A413283-250MG A413283-50MG A413283-5G
77155-746EA 36.05 USD
77155-746 77155-748 77155-750 77155-752 77572-284
2-Fluoro-4-nitrobenzenesulfonyl chloride 97%
2-Fluoro-4-nitrobenzenesulfonyl chloride

Precaución: for Research Use Only

Formula: C₆H₃ClFNO₄S
MW: 239.609 g/mol
MDL Number: MFCD11858095
CAS Number: 1146290-36-9
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR