Methyl 6-chloro-1H-indole-2-carboxylate 98%

Supplier: Ambeed
Warning

A412487-1G A412487-250MG A412487-5G
77155-734EA 101.97 USD
77155-734 77155-736 77155-738
Methyl 6-chloro-1H-indole-2-carboxylate 98%
Methyl 6-chloro-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈ClNO₂
MW: 209.62 g/mol
MDL Number: MFCD04966992
CAS Number: 98081-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR