3-Bromo-1-phenyl-5-(pyridin-2-yl)-1,2-dihydropyridin-2-one 95%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-(2-pyridyl)-1-phenyl-1,2-dihydropyridin-2-one

A412200-1G A412200-250MG A412200-5G
77155-728EA 80.34 USD
77155-728 77155-730 77155-732
3-Bromo-1-phenyl-5-(pyridin-2-yl)-1,2-dihydropyridin-2-one 95%
3-Bromo-1-phenyl-5-(pyridin-2-yl)-1,2-dihydropyridin-2-one

Precaución: for Research Use Only

Formula: C₁₆H₁₁BrN₂O
MW: 327.175 g/mol
MDL Number: MFCD21607392
CAS Number: 381248-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR