Ruthenium(III) chloride oxide 98%

Supplier: Ambeed
Warning

Synonyms: Ruthenium oxychloride, Tetraamminechlorohydroxoruthenium(III) chloride, Ruthenium Red

A412164-1G A412164-1MG A412164-250MG
77155-722EA 147 USD
77155-722 77155-724 77155-726
Ruthenium(III) chloride oxide 98%
Ruthenium(III) chloride oxide

Precaución: for Research Use Only

Formula: H₄₂Cl₆N₁₄O₂Ru₃
MW: 786.354 g/mol
MDL Number: MFCD00011479
CAS Number: 11103-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR