γ-Cyclodextrine hydrate 99%

Supplier: Ambeed
Warning

Synonyms: γ-Cyclodextrin hydrate, γ-Cyclodextrin xhydrate

A411212-10G A411212-25G A411212-100G A411212-500G
77155-720EA 18.54 USD
77155-720 77613-254 77686-133 77686-134
γ-Cyclodextrine hydrate 99%
γ-Cyclodextrine hydrate

Precaución: for Research Use Only

Formula: C₄₈H₈₀O₄₀·H₂O
MW: 1297.15 g/mol (anhy)
MDL Number: MFCD00149574
CAS Number: 91464-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR