(2R,4S)-rel-tert-Butyl 4-hydroxy-2-methylpiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A410758-1G A410758-250MG
77155-708EA 97.85 USD
77155-708 77155-710
(2R,4S)-rel-tert-Butyl 4-hydroxy-2-methylpiperidine-1-carboxylate 95%
(2R,4S)-rel-tert-Butyl 4-hydroxy-2-methylpiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₃
MW: 215.29 g/mol
MDL Number: MFCD28155048
CAS Number: 152491-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR