tert-Butyl 3,3-difluoropiperidin-4-ylcarbamate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (3,3-difluoropiperidin-4-yl)carbamate

A410491-100MG A410491-1G A410491-250MG
77155-700EA 61.8 USD
77155-700 77594-252 77686-132
tert-Butyl 3,3-difluoropiperidin-4-ylcarbamate 95%
tert-Butyl 3,3-difluoropiperidin-4-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₈F₂N₂O₂
MW: 236.259 g/mol
MDL Number: MFCD18791569
CAS Number: 1263180-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR