Methyl-1-methyl-4-nitro-1H-pyrazole-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-methyl-4-nitro-1H-pyrazole-5-carboxylate

A409867-10G A409867-1G A409867-250MG A409867-25G A409867-5G A409867-100G
77155-688EA 180.25 USD
77155-688 77155-690 77155-692 77155-694 77155-696 77686-131
Methyl-1-methyl-4-nitro-1H-pyrazole-5-carboxylate 98%
Methyl-1-methyl-4-nitro-1H-pyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇N₃O₄
MW: 185.14 g/mol
MDL Number: MFCD02110822
CAS Number: 309740-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR