2-(4-Fluorophenoxy)propionic acid 95%

Supplier: Ambeed
Warning

A409739-1G A409739-5G
77155-684EA 53 USD
77155-684 77155-686
2-(4-Fluorophenoxy)propionic acid 95%
2-(4-Fluorophenoxy)propionic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MDL Number: MFCD02295725
CAS Number: 2967-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR