Diisopropyl phthalate 98%

Supplier: Ambeed
Warning

Synonyms: Diisopropyl Phthalate

A409672-100G A409672-25G A409672-5G
77155-678EA 81 USD
77155-678 77155-680 77155-682
Diisopropyl phthalate 98%
Diisopropyl phthalate

Precaución: for Research Use Only

Formula: C₁₄H₁₈O₄
MW: 250.2903 g/mol
MDL Number: MFCD00053717
CAS Number: 605-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR