(S)-3-((tert-Butoxycarbonyl)amino)-2-(4-chlorophenyl)propanoic acid 95%
Supplier: Ambeed
|
Warning
|
A408889-100MG
A408889-1G
A408889-250MG
77155-664EA
78.28
USD
77155-664
77155-666
77155-668
(S)-3-((tert-Butoxycarbonyl)amino)-2-(4-chlorophenyl)propanoic acid 95%
(S)-3-((tert-Butoxycarbonyl)amino)-2-(4-chlorophenyl)propanoic acid
Precaución: for Research Use Only
Formula:
C₁₄H₁₈ClNO₄ MW: 299.75 g/mol |
MDL Number:
MFCD16394572 CAS Number: 1001180-04-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...