4-Hexen-3-one 98%

Supplier: Ambeed
Danger

A408077-5G A408077-25G
77155-654EA 17 USD
77155-654 77901-451
4-Hexen-3-one 98%
4-Hexen-3-one

Precaución: for Research Use Only

Formula: C₆H₁₀O
MDL Number: MFCD00010631
CAS Number: 2497-21-4
UN: 1224
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR