Methyl 4-Aminoindole-6-carboxylate 97%

Supplier: Ambeed
Warning

A407250-100MG A407250-250MG A407250-1G
77155-648EA 13 USD
77155-648 77155-650 77901-222
Methyl 4-Aminoindole-6-carboxylate 97%
Methyl 4-Aminoindole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MDL Number: MFCD00461176
CAS Number: 121561-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR