Ethyl 7-chloro-3-hydroxybenzo[b]thiophene-2-carboxylate 98%

Supplier: Ambeed
Warning

A404256-100MG A404256-1G A404256-250MG
77155-628EA 12 USD
77155-628 77155-630 77155-632
Ethyl 7-chloro-3-hydroxybenzo[b]thiophene-2-carboxylate 98%
Ethyl 7-chloro-3-hydroxybenzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉ClO₃S
MDL Number: MFCD29044877
CAS Number: 1825392-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR