4-Iodophenyl acetate 95+%

Supplier: Ambeed
Warning

A404005-1G A404005-250MG
77155-624EA 63.86 USD
77155-624 77155-626
4-Iodophenyl acetate 95+%
4-Iodophenyl acetate

Precaución: for Research Use Only

Formula: C₈H₇IO₂
MW: 262.04 g/mol
MDL Number: MFCD00748429
CAS Number: 33527-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR