3-((2-Chloro-4-nitrophenoxy)methyl)benzoic acid 95%

Supplier: Ambeed
Warning

A401777-100MG A401777-250MG
77155-616EA 15.45 USD
77155-616 77155-618
3-((2-Chloro-4-nitrophenoxy)methyl)benzoic acid 95%
3-((2-Chloro-4-nitrophenoxy)methyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀ClNO₅
MW: 307.69 g/mol
MDL Number: MFCD03421912
CAS Number: 30880-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR