Ethyl-N-(tert-butoxycarbonyl)-D-pyroglutamate 97%
Supplier: Ambeed
|
Warning
|
A400675-100G
A400675-10G
A400675-1G
A400675-25G
A400675-5G
A400675-500G
A400675-1000G
77155-600EA
66
USD
77155-600
77155-602
77155-604
77155-606
77155-608
77613-216
77901-868
Ethyl-N-(tert-butoxycarbonyl)-D-pyroglutamate 97%
Ethyl-N-(tert-butoxycarbonyl)-D-pyroglutamate
Precaución: for Research Use Only
Formula:
C₁₂H₁₉NO₅ |
MDL Number:
MFCD09261329 CAS Number: 144978-35-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...