N,N-Dimethylethylendiamine 99%

Supplier: Ambeed
Danger

Synonyms: N,N-Dimethylethylenediamine, asym. Dimethylethylendiamine, unsym-Dimethylethylenediamine, 2-(Dimethylamino)ethylamine, 1-Amino-2-dimethylaminoethane, 2-Dimethylaminoethylamine, 2-Aminoethyldimethylamine

A399439-25G A399439-100G
77155-594EA 6.18 USD
77155-594 77686-105
N,N-Dimethylethylendiamine 99%
N,N-Dimethylethylendiamine

Precaución: for Research Use Only

Formula: C₄H₁₂N₂
MW: 88.15 g/mol
MDL Number: MFCD00008175
CAS Number: 108-00-9
UN: 2733
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR