2-Thiazol-2-ylethylamine 98%

Supplier: Ambeed
Danger

Synonyms: 2-(Thiazol-2-yl)ethanamine

A398128-1G A398128-250MG A398128-5G
77155-584EA 151.41 USD
77155-584 77155-586 77155-588
2-Thiazol-2-ylethylamine 98%
2-Thiazol-2-ylethylamine

Precaución: for Research Use Only

Formula: C₅H₈N₂S
MW: 128.2 g/mol
MDL Number: MFCD08060802
CAS Number: 18453-07-1
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR