Allyltrimethoxysilane 97%

Supplier: Ambeed
Danger

Synonyms: Trimethoxyallylsilane

A397999-1G A397999-25G
77155-580EA 8.24 USD
77155-580 77155-582
Allyltrimethoxysilane 97%
Allyltrimethoxysilane

Precaución: for Research Use Only

Formula: H₂CCHCH₂Si(OCH₃)₃
MW: 162.26 g/mol
MDL Number: MFCD00053865
CAS Number: 2551-83-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR