Pazufloxacin mesylate 98%

Supplier: Ambeed
Warning

A395221-100MG A395221-25MG A395221-25G A395221-5G
77155-558EA 7 USD
77155-558 77155-560 77686-092 77686-093
Pazufloxacin mesylate 98%
Pazufloxacin mesylate

Precaución: for Research Use Only

Formula: C₁₆H₁₅FN₂O₄
MDL Number: MFCD00913262
CAS Number: 163680-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR