Benzyl 6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 3-Cbz-6-oxa-3-azabicyclo[3.1.0]hexane

A394707-1G A394707-5G
77155-550EA 26.78 USD
77155-550 77613-198
Benzyl 6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate 97%
Benzyl 6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₃
MW: 219.237 g/mol
MDL Number: MFCD09608058
CAS Number: 31865-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR