5-Bromo-1-pentene 98%

Supplier: Ambeed
Danger

A394665-10G A394665-1G A394665-25G A394665-5G A394665-100G
77155-542EA 8.24 USD
77155-542 77155-544 77155-546 77155-548 77594-204
5-Bromo-1-pentene 98%
5-Bromo-1-pentene

Precaución: for Research Use Only

Formula: C₅H₉Br
MW: 149.03 g/mol
MDL Number: MFCD00000264
CAS Number: 1119-51-3
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR