2-Chloro-6-fluorophenylisothiocyanate 95+%

Supplier: Ambeed
Danger

A394531-10G A394531-1G A394531-5G
77155-536EA 127 USD
77155-536 77155-538 77155-540
2-Chloro-6-fluorophenylisothiocyanate 95+%
2-Chloro-6-fluorophenylisothiocyanate

Precaución: for Research Use Only

Formula: C₇H₃ClFNS
MW: 187.62 g/mol
MDL Number: MFCD09878160
CAS Number: 899806-25-8
UN: 1760
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR